Orders LinkContactUsLinkComponent
Sample Vials, Clear And Amber, Electron Microscopy Sciences
Sample Vials, Clear And Amber, Electron Microscopy Sciences
Certificates
Sample Vials, Clear And Amber, Electron Microscopy Sciences
Sample Vials, Clear And Amber, Electron Microscopy Sciences
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Multi purpose glass sample vials suited for research studies. Clear vials are manufactured from Wheaton-33R a low extractable borosilicate glass which protects against changes in the pH and maintains sample integrity. Amber vials are manufactured from Wheaton '320' borosilicate glass and protects light-sensitive samples.
  • Economically priced
  • Come with lined caps
  • Can be sterilized
  • Can be stored at low temperatures
 : Packaged in two shrink-wrapped packs of 100 vials. Caps are packaged separately.
Product Family Options
Product Information
  • Capacity
    Diameter ×H
    Color
    Cap included
    Availability
    Price
  • 100500-262
    Sample Vial
    Sample Vial
    1.80 ml
    12×35 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $567.37
    $567.37
    Log in for availability
    Each
    Each - $567.37
    $567.37
    Case of 200
       $567.37
    Each
       $567.37
    Supplier Number: 60992-01
  • 100500-272
    Sample Vial
    Sample Vial
    1.50 ml
    12×35 mm
    Amber
    Yes
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $833.21
    $833.21
    Log in for availability
    Each
    Each - $833.21
    $833.21
    Case of 200
       $833.21
    Each
       $833.21
    Supplier Number: 60993-01
  • 100500-274
    Sample Vial
    Sample Vial
    2.00 ml
    12×38 mm
    Amber
    Yes
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $916.73
    $916.73
    Log in for availability
    Each
    Each - $916.73
    $916.73
    Case of 200
       $916.73
    Each
       $916.73
    Supplier Number: 60993-02
  • 100500-276
    Sample Vial
    Sample Vial
    4.00 ml
    15×48 mm
    Amber
    Yes
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $986.49
    $986.49
    Log in for availability
    Each
    Each - $986.49
    $986.49
    Case of 200
       $986.49
    Each
       $986.49
    Supplier Number: 60993-04
  • 100501-738
    Sample Vial
    Sample Vial
    4.00 ml
    15×48 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $720.36
    $720.36
    Log in for availability
    Each
    Each - $720.36
    $720.36
    Case of 200
       $720.36
    Each
       $720.36
    Supplier Number: 60992-04
  • 100500-278
    Sample Vial
    Sample Vial
    8.00 ml
    17×63 mm
    Amber
    Yes
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $1,054.16
    $1,054.16
    Log in for availability
    Each
    Each - $1,054.16
    $1,054.16
    Case of 200
       $1,054.16
    Each
       $1,054.16
    Supplier Number: 60993-08
  • 100503-122
    Sample Vial
    Sample Vial
    12.00 ml
    19×68 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $854.67
    $854.67
    Log in for availability
    Each
    Each - $854.67
    $854.67
    Case of 200
       $854.67
    Each
       $854.67
    Supplier Number: 60992-12
  • 100500-268
    Sample Vial
    Sample Vial
    16.00 ml
    21×73 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $951.51
    $951.51
    Log in for availability
    Each
    Each - $951.51
    $951.51
    Case of 200
       $951.51
    Each
       $951.51
    Supplier Number: 60992-16
  • 100500-270
    Sample Vial
    Sample Vial
    24.00 ml
    23×88 mm
    Clear
    Yes
    Discontinued
    Supplier Number: 60992-24
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.