Orders LinkContactUsLinkComponent
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Certificates
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A large size storage set. It comes with square clear glass bottles (1 ounce capacity) and black screw caps with plastic linings.
Alpha-numerical indexing system allows quick identification and location of stored items.
Delivery information: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
Product Family Options
Product Information
  • Capacity
    Cap included
    Availability
    Price
  • 100502-370
    Square Bottle Series 4
    Square Bottle Series 4
    116.00 ml
    Yes
    Log in for availability
    Log in for availability
    Each
    Each - $325.11
    $325.11
    Each
       $325.11
    Supplier Number: 64283
  • 100496-188
    Square Bottle Series 2
    Square Bottle Series 2
    78.00 ml
    Yes
    Discontinued
    Supplier Number: 64282
  • 100496-190
    Square Bottle Series
    Square Bottle Series
    232.00 ml
    Yes
    Log in for availability
    Log in for availability
    Each
    Each - $285.47
    $285.47
    Each
       $285.47
    Supplier Number: 64284
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)