Sample Storage Sets; Snap Cap Shell Vial, Electron Microscopy Sciences
About this item
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom.
Alpha-numerical indexing system allows quick identification and location of stored items.
Ordering information: For vial sizes of 10 and 12 dram please call us for prices and availability.
Delivery information: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
1 Options Available Below
Product Details & Documents
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom.
Alpha-numerical indexing system allows quick identification and location of stored items.
Ordering information: For vial sizes of 10 and 12 dram please call us for prices and availability.
Delivery information: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
Product Family Options
Product Information
- CapacityCap includedCap typeAvailabilityPrice
- 100496-182Shell vial series, dram 4Supplier Number: 64277
- 100496-184Shell vial series, dram 7Shell vial series, dram 736.60 mlYesSnap capDiscontinued-Supplier Number: 64278
- 100496-180Shell vial series, dram 3Shell vial series, dram 310.40 mlYesSnap capDiscontinued-Supplier Number: 64276
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.