Orders LinkContactUsLinkComponent
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
Certificates
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These vials are for use with autosamplers that require 15×45 mm vials. The vials are available in either clear or amber glass or in natural polypropylene, without 15 mm snap plug cap.
  • Conical interior for maximum sample recovery
Product Family Options
Product Information
  • Capacity
    Diameter ×H
    Material
    Color
    Cap included
    Availability
    Price
  • 100489-484
    Shell Vials
    Shell Vials
    3.00 ml
    15×45 mm
    PP
    No
    Discontinued
    Supplier Number: -
  • 100502-742
    Shell Vials
    Shell Vials
    4.00 ml
    15×45 mm
    Glass
    Amber
    No
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $83.10
    $83.10
    Log in for availability
    Pack of 100
    Pack of 100 - $63.77
    $63.77
    Each
       $83.10
    Pack of 100
       $63.77
    Supplier Number: 72627-4A
  • 100489-488
    Shell Vials
    Shell Vials
    4.00 ml
    15×45 mm
    PP
    No
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $86.03
    $86.03
    Log in for availability
    Pack of 100
    Pack of 100 - $65.74
    $65.74
    Each
       $86.03
    Pack of 100
       $65.74
    Supplier Number: 72627-4P
  • 100489-486
    Shell Vials
    Shell Vials
    4.00 ml
    15×45 mm
    Glass
    Clear
    No
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $87.31
    $87.31
    Log in for availability
    Pack of 100
    Pack of 100 - $66.72
    $66.72
    Each
       $87.31
    Pack of 100
       $66.72
    Supplier Number: 72627-4C
Caps
Product Information
  • Catalog #
    Color
    Description
    Availability
    Price
  • Natural
    Snap Plug Cap for I.D.15 mm × O.D.45 mm
    Log in for availability
    Log in for availability
    Each
    Each - $58.42
    $58.42
    Log in for availability
    Pack of 100
    Pack of 100 - $58.42
    $58.42
    Each
       $58.42
    Pack of 100
       $58.42
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.