Rinzl Plastic Cover Slips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Made from clear vinyl plastic.
- Unbreakable, will not scratch or cut fingers
- Used in Biological and Medical labs, etc.
- Thickness 0.01" or 0.28 mm
- Refractive index of glass
- Economical and half the price of glass
- Not autofluorescent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.