Orders LinkContactUsLinkComponent
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
Certificates
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
CoverWell™ Perfusion Chambers, Electron Microscopy Sciences
13 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CoverWell™ Perfusion Chambers allows up to 16 different specimens on one coverglass.
  • Chambers are removable for permanent mounting of specimens
  • CoverWell™ chambers are made from medical grade silicone bonded to clear, UV transparent plastic
  • Available in multiple designs allowing up to sixteen different specimens or analytical test on one coverglass
  • Ideal for cell imaging on upright or inverted microscopes
"Press-to-Seal" covers form water-tight, multiwall cell incubation or cytochemistry chamber when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports (1.5 mm diameter) without disturbing or cross-contaminating specimens in adjacent wells.
Product Family Options
Product Information
  • Depth
    Availability
    Price
  • 100490-762
    1 well, 180 µL
    1 well, 180 µL
    0.6 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $349.22
    $349.22
    Log in for availability
    Pack of 40
    Pack of 40 - $289.41
    $289.41
    Each
       $349.22
    Pack of 40
       $289.41
    Supplier Number: 70326-10
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-762
    Minority or Woman-Owned Business Enterprise
  • 100490-768
    1 well, 200 µL
    1 well, 200 µL
    0.6 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $720.25
    $720.25
    Log in for availability
    Pack of 50
    Pack of 50 - $541.47
    $541.47
    Each
       $720.25
    Pack of 50
       $541.47
    Supplier Number: 70326-20
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-768
    Minority or Woman-Owned Business Enterprise
  • 100502-780
    1 well, 300 µL
    1 well, 300 µL
    0.9 mm
    Discontinued
    Supplier Number: 70326-12
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100502-780
    Minority or Woman-Owned Business Enterprise
  • 100490-770
    1 well, 350 µL
    1 well, 350 µL
    0.6 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $565.37
    $565.37
    Log in for availability
    Pack of 40
    Pack of 40 - $410.77
    $410.77
    Each
       $565.37
    Pack of 40
       $410.77
    Supplier Number: 70326-22
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-770
    Minority or Woman-Owned Business Enterprise
  • 100490-764
    1 well, 520 µL
    1 well, 520 µL
    1.7 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $295.91
    $295.91
    Log in for availability
    Pack of 20
    Pack of 20 - $225.12
    $225.12
    Each
       $295.91
    Pack of 20
       $225.12
    Supplier Number: 70326-14
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-764
    Minority or Woman-Owned Business Enterprise
  • 100501-300
    1 well, 550 µL
    1 well, 550 µL
    0.9 mm
    Discontinued
    Supplier Number: 70326-24
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100501-300
    Minority or Woman-Owned Business Enterprise
  • 100490-766
    1 well, 750 µL
    1 well, 750 µL
    2.5 mm
    Discontinued
    Supplier Number: 70326-16
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-766
    Minority or Woman-Owned Business Enterprise
  • 100490-772
    1 well, 1000 µL
    1 well, 1000 µL
    1.7 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $382.91
    $382.91
    Log in for availability
    Pack of 20
    Pack of 20 - $276.15
    $276.15
    Each
       $382.91
    Pack of 20
       $276.15
    Supplier Number: 70326-26
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-772
    Minority or Woman-Owned Business Enterprise
  • 100490-774
    1 well, 1550 µL
    1 well, 1550 µL
    2.5 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $382.91
    $382.91
    Log in for availability
    Pack of 20
    Pack of 20 - $276.15
    $276.15
    Each
       $382.91
    Pack of 20
       $276.15
    Supplier Number: 70326-28
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-774
    Minority or Woman-Owned Business Enterprise
  • 100501-302
    3 well, 120 µL
    3 well, 120 µL
    0.9 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $429.34
    $429.34
    Log in for availability
    Pack of 40
    Pack of 40 - $312.75
    $312.75
    Each
       $429.34
    Pack of 40
       $312.75
    Supplier Number: 70326-32
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100501-302
    Minority or Woman-Owned Business Enterprise
  • 100490-778
    3 well, 200 µL
    3 well, 200 µL
    1.7 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $381.52
    $381.52
    Log in for availability
    Pack of 20
    Pack of 20 - $280.07
    $280.07
    Each
       $381.52
    Pack of 20
       $280.07
    Supplier Number: 70326-34
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-778
    Minority or Woman-Owned Business Enterprise
  • 100490-780
    3 well, 300 µL
    3 well, 300 µL
    2.5 mm
    Discontinued
    Supplier Number: 70326-36
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-780
    Minority or Woman-Owned Business Enterprise
  • 100490-782
    4 well, 70 µL
    4 well, 70 µL
    0.6 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $640.89
    $640.89
    Log in for availability
    Pack of 40
    Pack of 40 - $464.44
    $464.44
    Each
       $640.89
    Pack of 40
       $464.44
    Supplier Number: 70326-40
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-782
    Minority or Woman-Owned Business Enterprise
  • 100490-784
    4 well, 110 µL
    4 well, 110 µL
    0.9 mm
    Discontinued
    Supplier Number: 70326-42
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-784
    Minority or Woman-Owned Business Enterprise
  • 100502-782
    4 well, 300 µL
    4 well, 300 µL
    2.5 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $447.00
    $447.00
    Log in for availability
    Pack of 20
    Pack of 20 - $323.77
    $323.77
    Each
       $447.00
    Pack of 20
       $323.77
    Supplier Number: 70326-46
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100502-782
    Minority or Woman-Owned Business Enterprise
  • 100490-788
    8 well, 35 µL
    8 well, 35 µL
    0.6 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $379.18
    $379.18
    Log in for availability
    Pack of 20
    Pack of 20 - $282.41
    $282.41
    Each
       $379.18
    Pack of 20
       $282.41
    Supplier Number: 70326-50
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-788
    Minority or Woman-Owned Business Enterprise
  • 100490-790
    8 well, 60 µL
    8 well, 60 µL
    0.9 mm
    Discontinued
    Supplier Number: 70326-52
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-790
    Minority or Woman-Owned Business Enterprise
  • 100490-792
    8 well, 100 µL
    8 well, 100 µL
    1.7 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $311.23
    $311.23
    Log in for availability
    Pack of 10
    Pack of 10 - $227.99
    $227.99
    Each
       $311.23
    Pack of 10
       $227.99
    Supplier Number: 70326-54
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-792
    Minority or Woman-Owned Business Enterprise
  • 100490-794
    8 well, 150 µL
    8 well, 150 µL
    2.5 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $327.03
    $327.03
    Log in for availability
    Pack of 10
    Pack of 10 - $237.08
    $237.08
    Each
       $327.03
    Pack of 10
       $237.08
    Supplier Number: 70326-56
    Specifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-794
    Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.