ContactUsLinkComponent
Parasite Magnifier, Electron Microscopy Sciences
Parasite Magnifier, Electron Microscopy Sciences
  100491-434
Minority or Woman-Owned Business Enterprise
undefined
Parasite Magnifier, Electron Microscopy Sciences
  100491-434
 :  68110
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Magnification:
    2.5×, 5×
  • Description:
    Parasite Magnifier
  • Cat. no.:
    100491-434
  • Supplier No.:
    68110
An improved version of the conventional magnifying glass.
A 20 mm diameter parasite-like lens is cemented on a large 75 mm diameter lens. Magnification of the large one is 2.5X, while the small lens area has the magnification of 5X. The magnified image is very sharp and clear. Weight: 111g