Paramat, Solidification point: 56-58°C embedding agent for histology
Catalog # 100496-598
Supplier: Electron Microscopy Sciences
Minority or Woman-Owned Business Enterprise
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:10 kg
- Cat. no.:100496-598
- Supplier No.:19286-10
Specifications
Specification Test Results
- Appearance:Milky-white, pastilles
- Crystalline Structure under the microscope:Needles & Platelets
- Effect of heating (@60°C/100g):Clear melt within max. 16 hrs
- Solubility in Xylene (@20°C/1g in 50 ml):Clear solution within max. 16 hrs
- Cutting results for 4*m sections:Over 75% usable single; over 65% usable serial
- Solubility in Xylene (section):Max. 45 sec
- Melting Point:56-58°C
- Solidification Point:56-58°C
- Tears and Vascuoles:None
- Adhesion of sections Very good; protein, to microscope slides:glycerol etc. is not required to hold the section in place
- Free acid content:Max. 0.01%
- Block Structure:No mottling, no shrinking
- Block Solidification (room Temperature):About 2 hrs
Specification Test Results
About this item
:
:GHS07
CAS Number:8002-74-2
:8012-95-1