Specifications
- Description:Para-Marker
- Supplier No.:71313-20
Specifications
About this item
A wide tipped pen, designed for use with paraffin sections
The pen provides a sticky membrane to the glass slide which not only prevents paraffin sections from falling off but also keeps the sections flat on the slide.