Orders LinkContactUsLinkComponent
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Certificates
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Vannas micro scissors have extra fine tips and are 3" (75 mm) long. Available with a 3 mm or 5 mm cutting edge.
Product Family Options
Product Information
  • Blade length
    Blade type
    Availability
    Price
  • 100493-880
    Vannas Scissors Straight, Delicate
    Vannas Scissors Straight, Delicate
    3 mm (⅛")
    Extra fine
    Log in for availability
    Log in for availability
    Each
    Each - $993.40
    $993.40
    Each
       $993.40
    Supplier Number: 72932-01
  • 100493-882
    Vannas Scissors Curved, Delicate
    Vannas Scissors Curved, Delicate
    3 mm (⅛")
    Extra fine
    Log in for availability
    Log in for availability
    Each
    Each - $993.40
    $993.40
    Each
       $993.40
    Supplier Number: 72932-02
  • 100493-884
    Vannas Scissors Angle Forward, Delicate
    Vannas Scissors Angle Forward, Delicate
    3 mm (⅛")
    Extra fine
    Log in for availability
    Log in for availability
    Each
    Each - $851.12
    $851.12
    Each
       $851.12
    Supplier Number: 72932-03
  • 100493-886
    Vannas Scissors Angle On Side, Delicate
    Vannas Scissors Angle On Side, Delicate
    3 mm (⅛")
    Extra fine
    Log in for availability
    Log in for availability
    Each
    Each - $851.12
    $851.12
    Each
       $851.12
    Supplier Number: 72932-04
  • 100493-888
    Vannas Scissors Straight, Delicate
    Vannas Scissors Straight, Delicate
    5 mm
    Extra fine
    Log in for availability
    Log in for availability
    Each
    Each - $974.16
    $974.16
    Each
       $974.16
    Supplier Number: 72933-01
  • 100493-890
    Vannas Scissors Curved, Delicate
    Vannas Scissors Curved, Delicate
    5 mm
    Extra fine
    Log in for availability
    Log in for availability
    Each
    Each - $818.55
    $818.55
    Each
       $818.55
    Supplier Number: 72933-02
  • 100493-892
    Vannas Scissors Angle Forward, Delicate
    Vannas Scissors Angle Forward, Delicate
    5 mm
    Extra fine
    Log in for availability
    Log in for availability
    Each
    Each - $829.67
    $829.67
    Each
       $829.67
    Supplier Number: 72933-03
  • 100493-894
    Vannas Scissors Angle On Side, Delicate
    Vannas Scissors Angle On Side, Delicate
    5 mm
    Extra fine
    Log in for availability
    Log in for availability
    Each
    Each - $829.67
    $829.67
    Each
       $829.67
    Supplier Number: 72933-04
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)