Orders LinkContactUsLinkComponent
Mini Rotator, Electron Microscopy Sciences
Mini Rotator, Electron Microscopy Sciences
Mini Rotator, Electron Microscopy Sciences
Certificates
Mini Rotator, Electron Microscopy Sciences
Mini Rotator, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
It is heavy duty and has a permanent-magnet motor which maintains constant speed with a variable speed control between 2-80 rpm and its mixing action is able to be adjusted from a gentle slosh to a rapid agitation
The mini-rotator is a small, compact, rotator which is ideal for use on a lab bench; in a small incubator or refrigerator; in a biological cabinet; or in a fume hood or any other place where space is at a premium. It handles both large and small applications for micro centrifuge tubes and micro test tubes.
It is heavy duty and has a permanent-magnet motor which maintains constant speed with a variable speed control between 2-80 rpm and its mixing action is able to be adjusted from a gentle slosh to a rapid agitation. For optimum mixing, the chemical resistant cabinet is fixed at a 20 degree angle. Unit comes complete with an aluminum disk head with spring clips to hold 15 each for both 5-10 mm and 12-14 mm tubes. An option of 30 position tissue culture tube head is available.
For optimum mixing, the chemical resistant cabinet is fixed at a 20 degree angle. Unit comes complete with an aluminum disk head with spring clips to hold 15 each for both 5-10 mm and 12-14 mm tubes. An option of 30 position tissue culture tube head is available.
Product Family Options
Product Information
  • Electrical
    Availability
    Price
  • 100498-876
    Mini Rotator
    Mini Rotator
    120 V
    Log in for availability
    Log in for availability
    Each
    Each - $2,849.61
    $2,849.61
    Each
       $2,849.61
    Supplier Number: 65100-10
  • 100498-878
    Mini Rotator
    Mini Rotator
    220 V
    Log in for availability
    Log in for availability
    Each
    Each - $2,606.13
    $2,606.13
    Each
       $2,606.13
    Supplier Number: 65100-20
  • 100501-712
    Tissue Culture Head With Clips
    Tissue Culture Head With Clips
    Log in for availability
    Log in for availability
    Each
    Each - $1,042.30
    $1,042.30
    Each
       $1,042.30
    Supplier Number: 65100-50
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)