Orders LinkContactUsLinkComponent
Micro-Test Staining Dish, Electron Microscopy Sciences
Micro-Test Staining Dish, Electron Microscopy Sciences
Micro-Test/Staining Dish
Catalog # 100491-792
Supplier #:  71564
Micro-Test Staining Dish, Electron Microscopy Sciences
Catalog # 100491-792
Micro-Test Staining Dish, Electron Microscopy Sciences
Micro-Test Staining Dish, Electron Microscopy Sciences
Micro-Test/Staining Dish
Price
$159.39
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This staining dish is made from clear glass and has 10 cells in 2 rows of 5 each. Each cell is 2 mm deep and holds 0.15 ml of solution.
Very useful in specimen preparation, EM staining, and Boerner-Jones-Lukens microfluocculation test.
Measures 108 X 57 mm (4¼" x 2¼").
Specifications
  • Description:
    Micro-Test/Staining Dish
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.