ContactUsLinkComponent
Micro Sieve Set, Electron Microscopy Sciences
Micro Sieve Set, Electron Microscopy Sciences
Catalog # 100496-826
Minority or Woman-Owned Business Enterprise
undefined
Micro Sieve Set, Electron Microscopy Sciences
Catalog # 100496-826
Supplier Number:  50111-00
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Replacement Mesh Inserts, Set of 8
  • Cat. no.:
    100496-826
  • Supplier No.:
    50111-00
Specifications
About this item
This micro-sieve set consists of a stack of four interchangeable screen-holding sections, a catch pan, and a friction-fit cover. All the parts of the set are very easy to take apart for changing the mesh insert or for cleaning.
  • Entire unit is moulded of poly-propylene
  • Includes phosphor-bronzed mesh inserts in 25, 35, 45, 60, 80, 120, 170, and 230 mesh
  • Holding sections are 2" I.Ø and 6" height