Orders LinkContactUsLinkComponent
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Certificates
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
16 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tough-Spots® Labels are pre-cut peel-off labels specially for microcentrifuge tube tops. This chemically resistant, flexible labels that can withstand boiling, freezing to −196 °C, sustained humidity.
  • Polyolefin labels resist most organic solvents and caustic agents
  • They adhere to most plastics, glass and metals without cracking, peeling, or degrading
  • Heat-resistant, stay-flat sheets will not jam in laser printers
  • Label sheets are top-coated to readily accept laser printing
  • Assortment includes four sheets of blue, green, red, yellow and orange color
Made of polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
Product Family Options
Product Information
  • Color
    Diameter
    Packaging
    Availability
    Price
  • 101413-968
    Adhesive Tough-Spots on laser sheets, round
    Adhesive Tough-Spots on laser sheets, round
    Assorted
    3/8"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $209.27
    $209.27
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $177.56
    $177.56
    Each
       $209.27
    Pack of 3,840
       $177.56
    Supplier Number: 77585-05
  • 101413-970
    Adhesive Tough-Spots on laser sheets, round
    Adhesive Tough-Spots on laser sheets, round
    Blue
    3/8"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $218.42
    $218.42
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
    Supplier Number: 77585-B
  • 101413-972
    Adhesive Tough-Spots on laser sheets, round
    Adhesive Tough-Spots on laser sheets, round
    Green
    3/8"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $218.42
    $218.42
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
    Supplier Number: 77585-G
  • 101413-974
    Adhesive Tough-Spots on laser sheets, round
    Adhesive Tough-Spots on laser sheets, round
    Orange
    3/8"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $218.42
    $218.42
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
    Supplier Number: 77585-O
  • 101413-976
    Adhesive Tough-Spots on laser sheets, round
    Adhesive Tough-Spots on laser sheets, round
    Red
    3/8"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $218.42
    $218.42
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
    Supplier Number: 77585-R
  • 101413-978
    Adhesive Tough-Spots on laser sheets, round
    Adhesive Tough-Spots on laser sheets, round
    White
    3/8"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $210.25
    $210.25
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $210.25
    Pack of 3,840
       $178.38
    Supplier Number: 77585-W
  • 101413-980
    Adhesive Tough-Spots on laser sheets, round
    Adhesive Tough-Spots on laser sheets, round
    Yellow
    3/8"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $240.60
    $240.60
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $275.50
    $275.50
    Each
       $240.60
    Pack of 3,840
       $275.50
    Supplier Number: 77585-Y
  • 102098-604
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    White
    7/16"
    4680 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $217.44
    $217.44
    Log in for availability
    Pack of 4,680
    Pack of 4,680 - $177.56
    $177.56
    Each
       $217.44
    Pack of 4,680
       $177.56
    Supplier Number: 77585-44
  • 102098-614
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    Assorted
    1/2"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $217.44
    $217.44
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $177.56
    $177.56
    Each
       $217.44
    Pack of 3,840
       $177.56
    Supplier Number: 77587-05
  • 102098-616
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    Blue
    1/2"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $218.42
    $218.42
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
    Supplier Number: 77587-B
  • 102098-618
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    Green
    1/2"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $218.42
    $218.42
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
    Supplier Number: 77587-G
  • 102098-620
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    Orange
    1/2"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $218.42
    $218.42
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
    Supplier Number: 77587-O
  • 102098-622
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    Red
    1/2"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $218.42
    $218.42
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $178.38
    $178.38
    Each
       $218.42
    Pack of 3,840
       $178.38
    Supplier Number: 77587-R
  • 102098-624
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    White
    1/2"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $217.44
    $217.44
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $177.56
    $177.56
    Each
       $217.44
    Pack of 3,840
       $177.56
    Supplier Number: 77587-W
  • 102098-626
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    Yellow
    1/2"
    3840 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $240.60
    $240.60
    Log in for availability
    Pack of 3,840
    Pack of 3,840 - $275.50
    $275.50
    Each
       $240.60
    Pack of 3,840
       $275.50
    Supplier Number: 77587-Y
  • 102098-630
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    White
    3/4"
    2160 labels/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $215.38
    $215.38
    Log in for availability
    Pack of 2,160
    Pack of 2,160 - $175.89
    $175.89
    Each
       $215.38
    Pack of 2,160
       $175.89
    Supplier Number: 77588-75
  • 102098-628
    Adhesive laser labels, Tough-Spots, round
    Adhesive laser labels, Tough-Spots, round
    White
    1"
    1260 labels/pack
    Discontinued
    Supplier Number: 77588-100
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.