Wooden Picks, Electron Microscopy Sciences
Catalog # 100501-832
Supplier: Electron Microscopy Sciences
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Wood Picks
- Cat. no.:100501-832
- Supplier No.:72340
Specifications
About this item
They are round and double tapered; used for cleaning pole pieces and parts of the EM. Also used for orienting the specimen during the embedding stage.