Orders LinkContactUsLinkComponent
Jaw Forceps, Electron Microscopy Sciences
Jaw Forceps, Electron Microscopy Sciences
8" Jaw Forceps
Catalog # 100492-026
Supplier #:  72925
Jaw Forceps, Electron Microscopy Sciences
Catalog # 100492-026
Jaw Forceps, Electron Microscopy Sciences
Jaw Forceps, Electron Microscopy Sciences
8" Jaw Forceps
Price
$37.68
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel jaw forceps with a guide pin. Curved, serrated tips.
Specifications
  • Description:
    8" Jaw Forceps
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)