Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
About this item
Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation.
- Consists of a threaded handle (MT1), 4.75" long and an interchangeable threaded base with tool tip
- Available in 32 different tip configurations with interchangeable handles
- Tip material is hardened tool steel unless noted
36 Options Available Below
Product Details & Documents
Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation.
- Consists of a threaded handle (MT1), 4.75" long and an interchangeable threaded base with tool tip
- Available in 32 different tip configurations with interchangeable handles
- Tip material is hardened tool steel unless noted
Product Family Options
Product Information
- SizeTip configurationAvailabilityPrice
- 100497-686Micro-BurnisherMicro-Burnisher0.50 mmTip 19Discontinued-Supplier Number: 62091-19
- 100497-718Micro-Carbide ScriberMicro-Carbide Scriber0.25 mmTip 28Discontinued-Supplier Number: 62091-28-25
- 100501-662Micro-Carbide ScriberSupplier Number: 62091-28-50
- 100497-642Micro-ChiselMicro-Chisel0.12 mmTip 3Discontinued-Supplier Number: -
- 100501-628Micro-ChiselSupplier Number: 62091-03-50
- 100497-678Micro-DiamondSupplier Number: 62091-15-25
- 100497-676Micro-DiamondSupplier Number: 62091-15-50
- 100497-726Micro-Diamond FileSupplier Number: 62091-29-25
- 100497-648Micro-ForkSupplier Number: 62091-05-25
- 100501-636Micro-ForkSupplier Number: 62091-05-50
- 100497-636Micro-GraverSupplier Number: 62091-02-50
- 100497-672Micro-GraverMicro-Graver0.12 mmTip 13Discontinued-Supplier Number: 62091-13-0.12
- 100501-644Micro-GraverSupplier Number: 62091-13-50
- 100501-648Micro-Hook 90°Supplier Number: 62091-18-50
- 102093-934Micro-Hook 180°Micro-Hook 180°0.50 mmTip 20Discontinued-Supplier Number: 62091-20-50
- 102093-932Micro-Hook 180°Supplier Number: 62091-20-25
- 100497-732Micro-HookSupplier Number: 62091-32
- 100497-654Micro-Knife 20°Supplier Number: 62091-08-25
- 100501-638Micro-Knife 20°Micro-Knife 20°0.50 mmTip 8Discontinued-Supplier Number: 62091-08-50
- 100501-642Micro-Knife 45°Micro-Knife 45°0.50 mmTip 9Discontinued-Supplier Number: 62091-09-50
- 100497-730Micro-LoopMicro-Loop0.50 mmTip 31Discontinued-Supplier Number: 62091-31
- 100497-650Micro-MirrorMicro-Mirror0.50 mmTip 6Discontinued-Supplier Number: 62091-06
- 100497-664Micro-MirrorSupplier Number: 62091-10
- 100497-632Micro-NeedleSupplier Number: 62091-01-12
- 100497-630Micro-NeedleSupplier Number: 62091-01-25
- 100497-628Micro-NeedleSupplier Number: 62091-01-50
- 102093-938Micro-NeedleMicro-Needle0.025 mmTip 24Discontinued-Supplier Number: -
- 100497-700Micro-NeedleSupplier Number: 62091-24-12
- 100501-656Micro-NeedleSupplier Number: 62091-24-25
- 100497-698Micro-NeedleSupplier Number: 62091-24-50
- 100497-668Micro-Prober 45°Supplier Number: 62091-12-25
- 100497-666Micro-Prober 90°Supplier Number: 62091-11-25
- 100502-470Micro-Prober 90°Supplier Number: 62091-11-50
- 100501-654Micro-SawMicro-Saw0.50 mmTip 22Discontinued-Supplier Number: 62091-22
- 100497-652Micro-ScaleSupplier Number: 62091-07-0.50
- 102093-924Micro-SpadeSupplier Number: 62091-04-025
- 100497-646Micro-SpadeSupplier Number: 62091-04-12
- 100501-632Micro-SpadeSupplier Number: 62091-04-50
- 100497-680Micro-SpatulaSupplier Number: 62091-16-0.50
- 100497-706Ultra Micro-NeedleSupplier Number: 62091-25-12
- 100497-710Ultra Micro-NeedleSupplier Number: 62091-26-12
- 100497-704Ultra-NeedleSupplier Number: 62091-25-25
- 100501-658Ultra-NeedleSupplier Number: 62091-25-50
- 100497-708Ultra-NeedleSupplier Number: 62091-26-25
- 100501-660Ultra-NeedleUltra-Needle0.50 mmTip 26Discontinued-Supplier Number: 62091-26-50
- 100497-716Ultra-NeedleSupplier Number: 62091-27-12
- 100497-712Ultra-NeedleSupplier Number: 62091-27-50
- 100497-626Handle Mt1, Gold AnodizedSupplier Number: 62090-00
- 100497-714Ultra-NeedleUltra-Needle0.25 mmTip 27Discontinued-Supplier Number: 62091-27-25
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



