ContactUsLinkComponent
Hardened Steel Deburring Tool Set, Electron Microscopy Sciences
Hardened Steel Deburring Tool Set, Electron Microscopy Sciences
Catalog # 100497-998
Minority or Woman-Owned Business Enterprise
undefined
Hardened Steel Deburring Tool Set, Electron Microscopy Sciences
Catalog # 100497-998
Supplier Number:  62161-SD
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Hardened Steel Deburring Tool Set
  • Cat. no.:
    100497-998
  • Supplier No.:
    62161-SD
Specifications
About this item
Made from hardened steel, this tool has three razor sharp cutting edges ranging from a very sharp point to one that is approximately 10 mm in length.
These nickel plated triangular tools are mounted in red and black plastic handles. Tool length is 30 mm by 2.1 mm across (1.2×0.08"). The overall length of the tool is 154 mm (5.7"). Two tools/set.