Orders LinkContactUsLinkComponent
Grid Coating Pen For TEM, Coat Quick G, Electron Microscopy Sciences
Grid Coating Pen For TEM, Coat Quick G, Electron Microscopy Sciences
Grid Coating Pen
Catalog # 100491-640
Supplier #:  70624
Grid Coating Pen For TEM, Coat Quick G, Electron Microscopy Sciences
Catalog # 100491-640
Grid Coating Pen For TEM, Coat Quick G, Electron Microscopy Sciences
Grid Coating Pen For TEM, Coat Quick G, Electron Microscopy Sciences
Grid Coating Pen
Price
$98.25
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Coat-Quick "G" pen improves the adherence of tissue sections onto the grids.
  • Used in pretreating grids prior to mounting supporting films such as formvar and carbon
  • Minimizes dislodging, widening, or breaking of the support film
With a touch of the pen to the grid, a thin layer of coating is applied to the grid. Drying takes place in approximately 1 to 2 minutes at room temperature. After it has dried the grid is ready for tissue mounting. The pen is also used in pretreating grids prior to mounting supporting films such as formvar and carbon; it minimizes dislodging, widening, or breaking of the support film.
Specifications
  • Description:
    Grid Coating Pen
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)