Orders LinkContactUsLinkComponent
Graduated Pitchers, Electron Microscopy Sciences
Graduated Pitchers, Electron Microscopy Sciences
Graduated Pitchers, Electron Microscopy Sciences
Certificates
Graduated Pitchers, Electron Microscopy Sciences
Graduated Pitchers, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Gracefully proportioned, molded pitchers with large comfortable handles and well designed pouring spouts.
  • Metric graduations, and crystal clear to the eye (polymethylpentene)
  • Resists heat to 175°C (374°F)
  • Shatter-proof
  • Excellent chemical resistance
Ideal for preparation of photographic chemicals.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 1000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $44.85
    $44.85
    Each
       $44.85
    Supplier Number: 74370
  • 3000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $69.90
    $69.90
    Each
       $69.90
    Supplier Number: 74372
  • 2000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $54.39
    $54.39
    Each
       $54.39
    Supplier Number: 74371
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)