Orders LinkContactUsLinkComponent
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Certificates
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Product Family Options
Product Information
  • Capacity
    Size
    Availability
    Price
  • 100489-766
    With Plastic Snap Cap
    With Plastic Snap Cap
    32 ml
    32 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,008.16
    $1,008.16
    Log in for availability
    Pack of 144
    Pack of 144 - $768.65
    $768.65
    Each
       $1,008.16
    Pack of 144
       $768.65
    Supplier Number: 64260
  • 100488-918
    Replacement Snap Cap
    Replacement Snap Cap
    22 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $95.37
    $95.37
    Log in for availability
    Pack of 200
    Pack of 200 - $83.30
    $83.30
    Each
       $95.37
    Pack of 200
       $83.30
    Supplier Number: 64251-22
  • 100489-756
    With Plastic Snap Cap
    With Plastic Snap Cap
    4 ml
    4 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $745.84
    $745.84
    Log in for availability
    Pack of 144
    Pack of 144 - $568.94
    $568.94
    Each
       $745.84
    Pack of 144
       $568.94
    Supplier Number: 64250
  • 100489-758
    With Plastic Snap Cap
    With Plastic Snap Cap
    8 ml
    8 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $581.50
    $581.50
    Log in for availability
    Pack of 144
    Pack of 144 - $466.26
    $466.26
    Each
       $581.50
    Pack of 144
       $466.26
    Supplier Number: 64252
  • 100489-760
    With Plastic Snap Cap
    With Plastic Snap Cap
    12 ml
    12 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $724.50
    $724.50
    Log in for availability
    Pack of 144
    Pack of 144 - $544.80
    $544.80
    Each
       $724.50
    Pack of 144
       $544.80
    Supplier Number: 64254
  • 100489-762
    With Plastic Snap Cap
    With Plastic Snap Cap
    16 ml
    16 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $536.99
    $536.99
    Log in for availability
    Pack of 144
    Pack of 144 - $488.11
    $488.11
    Each
       $536.99
    Pack of 144
       $488.11
    Supplier Number: 64256
  • 100489-764
    With Plastic Snap Cap
    With Plastic Snap Cap
    24 ml
    24 ml
    Discontinued
    Supplier Number: -
  • 102095-026
    Replacement Snap Cap
    Replacement Snap Cap
    30 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $138.47
    $138.47
    Log in for availability
    Pack of 200
    Pack of 200 - $104.66
    $104.66
    Each
       $138.47
    Pack of 200
       $104.66
    Supplier Number: 64257-30
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)