ContactUsLinkComponent
Fluorescent Ruler, Electron Microscopy Sciences
Fluorescent Ruler, Electron Microscopy Sciences
Catalog # 100492-722
Minority or Woman-Owned Business Enterprise
undefined
Fluorescent Ruler, Electron Microscopy Sciences
Catalog # 100492-722
Supplier Number:  68137-01
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Fluorescent ruler
  • Cat. no.:
    100492-722
  • Supplier No.:
    68137-01
Specifications
About this item
It is most useful for all fluorescent viewing and fluorescent photographic work.
Fluorescent Scale in inches and centimeters. This metric/english plastic ruler is printed with a pink phosphorescent ink that glows in the dark.The scale will fluoresce visibly upon illumination with ultraviolet light and serves as a measure of nucleic acid mobility. The scale is 0 to 6" with ¹/₁₆" increments on one edge and 0 to 15 cm with 1 mm increments on the other.