Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences
About this item
The Ellipso-Spoon® Sampler has a spoon end and a spatula end and is made from polished stainless steel.
Its unique shape means no more material loss. This utensil’s specially curved shape permits contact with all corners of bottles and containers; therefore, no loss of material occurs. Crystals, powders, granulated and solid substances, and even thick liquids are easily retrieved and never fall back into the container.
3 Options Available Below
Product Details & Documents
The Ellipso-Spoon® Sampler has a spoon end and a spatula end and is made from polished stainless steel.
Its unique shape means no more material loss. This utensil’s specially curved shape permits contact with all corners of bottles and containers; therefore, no loss of material occurs. Crystals, powders, granulated and solid substances, and even thick liquids are easily retrieved and never fall back into the container.
Product Family Options
Product Information
- LengthAvailabilityPrice
- 100501-466Bowl size 28×65 mmSupplier Number: 72984-30
- 100493-952Bowl size 15×15 mmSupplier Number: 72984-21
- 100493-950Bowl size 15×15 mmSupplier Number: 72984-15
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.