ContactUsLinkComponent
Dow Corning® Vacuum Grease, Electron Microscopy Sciences
Dow Corning® Vacuum Grease, Electron Microscopy Sciences
Catalog # 100504-358
Minority or Woman-Owned Business Enterprise
undefined
Dow Corning® Vacuum Grease, Electron Microscopy Sciences
Catalog # 100504-358
Supplier Number:  60705
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Vacuum Grease
  • Size:
    150 g
  • Cat. no.:
    100504-358
  • Supplier No.:
    60705
Specifications
About this item
A silicone lubricant for glass-to-glass joints and glass-to-rubber connections. Resistant to most chemicals and is inert and heat stable.