Disposable Glass Tubes, Electron Microscopy Sciences
About this item
Tubes are made from borosilicate glass, round bottom and plain top.
The tubes measuring 16 mm in outer diameter and 125 mm in length are Flint glass only.
Labeling area in five pastel colors. Handy fused-in color patches, accept any type of ink or pencil.
14 Options Available Below
Product Details & Documents
Tubes are made from borosilicate glass, round bottom and plain top.
The tubes measuring 16 mm in outer diameter and 125 mm in length are Flint glass only.
Labeling area in five pastel colors. Handy fused-in color patches, accept any type of ink or pencil.
Product Family Options
Product Information
- HeightØAvailabilityPrice
- 100490-612Yellow LabelYellow Label75 mm12 mmEach$577.31Pack of 1,000$440.43Supplier Number: 64635-Y
- 100490-620White LabelWhite Label100 mm13 mmEach$615.31Pack of 1,000$493.32Supplier Number: 64636-W
- 100490-616Green LabelGreen Label100 mm13 mmEach$549.01Pack of 1,000$418.98Supplier Number: 64636-G
- 100490-634Blue LabelBlue Label125 mm16 mmEach$689.62Pack of 1,000$526.09Supplier Number: 64638-B
- 100490-604Blue LabelBlue Label75 mm12 mmEach$582.79Pack of 1,000$444.60Supplier Number: 64635-B
- 100490-608Orange LabelOrange Label75 mm12 mmEach$513.69Pack of 1,000$391.89Supplier Number: 64635-O
- 100490-618Orange LabelOrange Label100 mm13 mmEach$549.01Pack of 1,000$418.98Supplier Number: 64636-O
- 100490-636Orange LabelOrange Label125 mm16 mmEach$579.86Pack of 1,000$526.09Supplier Number: 64638-O
- 100490-626Green LabelGreen Label100 mm16 mmDiscontinued-Supplier Number: 64637-G
- 100490-632Yellow LabelYellow Label100 mm16 mmEach$655.95Pack of 1,000$499.94Supplier Number: 64637-Y
- 100490-606Green LabelGreen Label75 mm12 mmEach$582.79Pack of 1,000$444.60Supplier Number: 64635-G
- 100490-614Blue LabelBlue Label100 mm13 mmDiscontinued-Supplier Number: 64636-B
- 100490-624Blue LabelBlue Label100 mm16 mmEach$551.03Pack of 1,000$499.94Supplier Number: 64637-B
- 100490-610White LabelWhite Label75 mm12 mmEach$551.31Pack of 1,000$439.95Supplier Number: 64635-W
- 100490-622Yellow LabelYellow Label100 mm13 mmEach$629.63Pack of 1,000$479.80Supplier Number: 64636-Y
- 100490-630White LabelWhite Label100 mm16 mmEach$646.78Pack of 1,000$586.80Supplier Number: 64637-W
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)