Orders LinkContactUsLinkComponent
Crystalbond™, Electron Microscopy Sciences
Crystalbond™, Electron Microscopy Sciences
Crystalbond™, Electron Microscopy Sciences
Crystalbond™, Electron Microscopy Sciences
Temporary adhesives that can be washed away after machining.
  • Machining or slicing single crystal metal specimens
  • Grinding and polishing sapphire, ceramic, optical garnets, ferrites, and LCD glass
  • Dicing and slicing germanium and silicon wafers in semiconductor production
  • Dicing and slicing alumina and beryllia substrates for IC and microelectronic production
  • Holding beam leads in IC devices for pull-off tests
  • Dicing subminiature chip capacitors and microwave IC substrates
These wash away temporary adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing
Crystalbond™ 509:
Offers excellent adhesion to metals, glass, and ceramic. This material is the best for precise high purity work since it leaves no residue after dissolving and does not clog the diamond wheel as compared to conventional waxes. Each stick weighs about 90 grams.
Crystalbond™ 555:
The same as 509 but used in applications where it is desirable to have a temporary bond which is water soluble. Each stick weighs about 90 grams.
Crystalbond™ 590:
The same as 509 but offers maximum adhesion to subminiature parts. Each stick weighs about 225 grams.
Product Family Options
  • Description
    Size
  • Crystalbond 555
    90 g
    Each
    Each - $639.45
    $639.45
    Pack of 10
    Pack of 10 - $536.79
    $536.79
    Each
       $639.45
    Pack of 10
       $536.79
     : 50401-10
  • Crystalbond 590, 1 Stick
    225 g
    Each
    Each - $189.86
    $189.86
    Each
       $189.86
     : 50402-01
  • Crystalbond 509
    90 g
     : 50400-05
  • Crystal Bond Stripper-590, 100g
    100 g
    Each
    Each - $92.63
    $92.63
    Each
       $92.63
     : 50402-S
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)