Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
About this item
CYO-LOK vials are ideal for storage or transport of material of extremely low temperatures.
- Graduated in 0.5 ml increments
The vials have external threads, providing a smooth and uniform inner surface. This eliminates the problem of air born contamination and/or sample loss so common when vials with internal threads are used. The screw top incorporates a molded-in sealing ring, which assures a tight leak-proof seal.
1 Options Available Below
Product Details & Documents
CYO-LOK vials are ideal for storage or transport of material of extremely low temperatures.
- Graduated in 0.5 ml increments
The vials have external threads, providing a smooth and uniform inner surface. This eliminates the problem of air born contamination and/or sample loss so common when vials with internal threads are used. The screw top incorporates a molded-in sealing ring, which assures a tight leak-proof seal.
Product Family Options
Product Information
- CapacityCap includedSterilityAvailabilityPrice
- 2.50 mlYesNon sterileBox of 1,000$258.43Each$258.43Supplier Number: 72652
- Discontinued-Supplier Number: -
- Discontinued-Supplier Number: -
- 2.50 mlYesSterileDiscontinued-Supplier Number: -
- Discontinued-Supplier Number: -
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)