Cryo Labels, Electron Microscopy Sciences
About this item
A special formulated material for use at ultra-low temperatures and in cryo storage conditions, such as liquid nitrogen and oxygen.
- Will not peel or shrink
They are durable and adhere well to plastic as well as cardboard cryogenic storage boxes. Ten sheets of labels: five preprinted, five plain.
Packaging: Packaged as 20 labels/sheet.
1 Options Available Below
Product Details & Documents
A special formulated material for use at ultra-low temperatures and in cryo storage conditions, such as liquid nitrogen and oxygen.
- Will not peel or shrink
They are durable and adhere well to plastic as well as cardboard cryogenic storage boxes. Ten sheets of labels: five preprinted, five plain.
Packaging: Packaged as 20 labels/sheet.
Product Family Options
Product Information
- DimensionsPackagingAvailabilityPrice
- 100489-746Cryo labelsCryo labels25×50 mm200 labels/packEach$253.40Pack of 200$229.89Supplier Number: 62046
- 100496-126Cryo labelsCryo labels25×50 mm10 packs/lotDiscontinued-Supplier Number: 62047
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)