ContactUsLinkComponent
Cross Sectional Holder, Electron Microscopy Sciences
Cross Sectional Holder, Electron Microscopy Sciences
Catalog # 100494-348
Minority or Woman-Owned Business Enterprise
undefined
Cross Sectional Holder, Electron Microscopy Sciences
Catalog # 100494-348
Supplier Number:  75942-01
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Cross Sectional Holder
  • Cat. no.:
    100494-348
  • Supplier No.:
    75942-01
Specifications
About this item
Made from non-magnetic stainless steel with 3.1 mm (1/8") diameter pin and adjustable angle turn-screw.
Just insert specimens edge-on and observe the cross section directly.