Orders LinkContactUsLinkComponent
Cotton Tipped Applicator, Electron Microscopy Sciences
Cotton Tipped Applicator, Electron Microscopy Sciences
Cotton Tipped Applicator, Electron Microscopy Sciences
Certificates
Cotton Tipped Applicator, Electron Microscopy Sciences
Cotton Tipped Applicator, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A lint-free, 6" double cotton tipped applicator with one pointed end and one regular end on a 1/12" wooden shaft.
The pointed head size is 3/16" tapering to 1/32 x 5/8". Regular head size 3/16 x 5/8".
Packaging: Packed 100 sticks in a sealed bag.
Product Family Options
Product Information
  • Handle length
    Availability
    Price
  • 100489-876
    Double End, Tapered, Wooden Shaft
    Double End, Tapered, Wooden Shaft
    16 cm (6")
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $30.58
    $30.58
    Log in for availability
    Pack of 100
    Pack of 100 - $23.40
    $23.40
    Each
       $30.58
    Pack of 100
       $23.40
    Supplier Number: 72308
  • 100499-646
    Double End, Wooden Shaft
    Double End, Wooden Shaft
    16 cm (6")
    Log in for availability
    Log in for availability
    Each
    Each - $129.21
    $129.21
    Each
       $129.21
    Supplier Number: 72308-05
  • 100488-464
    Large Tip, Wooden Shaft
    Large Tip, Wooden Shaft
    16 cm (6")
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $40.90
    $40.90
    Log in for availability
    Pack of 5
    Pack of 5 - $31.76
    $31.76
    Each
       $40.90
    Pack of 5
       $31.76
    Supplier Number: 72309
  • 100496-338
    Anti-Static, Polypropylene Shaft
    Anti-Static, Polypropylene Shaft
    16 cm (6")
    Log in for availability
    Log in for availability
    Each
    Each - $109.68
    $109.68
    Each
       $109.68
    Supplier Number: 72306
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)