Concentric Circles Graticle, Electron Microscopy Sciences
About this item
For calibrated measurement in a light microscope.
- Scales in mm and inches available
- Surface chrome image
- Extended size range
The microscope graticle can be used for two-way measurement when calibrated as a micrometer.
14 Options Available Below
Product Details & Documents
For calibrated measurement in a light microscope.
- Scales in mm and inches available
- Surface chrome image
- Extended size range
The microscope graticle can be used for two-way measurement when calibrated as a micrometer.
Product Family Options
Product Information
- SizeSurfaceAvailabilityPrice
- 100499-080Graticle NE43Supplier Number: 67997-16
- 100499-086Graticle NE44Supplier Number: 67998-16
- 100499-092Graticle NE47Supplier Number: 67999-16
- 100499-200Graticle NE42Supplier Number: 68019-16
- 100499-094Graticle NE47Supplier Number: 67999-19
- 100499-202Graticle NE42Supplier Number: 68019-19
- 103305-416Graticle NE44Supplier Number: 67998-20.4
- 100499-084Graticle NE43Supplier Number: 67997-21
- 100499-090Graticle NE44Supplier Number: 67998-21
- 103305-418Graticle NE44Supplier Number: 67998-23
- 103305-420Graticle NE44Supplier Number: 67998-24.5
- 103305-422Graticle NE44Supplier Number: 67998-25
- 103305-424Graticle NE44Supplier Number: 67998-26
- 103305-426Graticle NE44Supplier Number: 67998-27
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)