Circular Cover Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Circular Cover Glass is made from pre-selected, pre-cleaned silicate glass.
- Thickness of #1 (0.13 to 0.17 mm), #1.5 (0.16 to 0.19 mm) and #2 (0.17 to 0.25 mm)
- Select from diameters of 12, 15, 18, 22, and 25 mm
Packaged with desiccants in a lint free box. Both cover glass cases and slide boxes convert to convenient slide storage files.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.