Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
About this item
Made from translucent polypropylene.
- Resistant to breakage
- Blue graduations, ring mark and inscriptions
Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.
NOTE: Exposure to temperatures above 60 °C may cause volume changes.
7 Options Available Below
Product Details & Documents
Made from translucent polypropylene.
- Resistant to breakage
- Blue graduations, ring mark and inscriptions
Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.
NOTE: Exposure to temperatures above 60 °C may cause volume changes.
Product Family Options
Product Information
- CapacityAvailabilityPrice
- 100489-204VolumetricVolumetric10 mlEach$364.67Pack of 3$263.16Supplier Number: 70952-10
- 100489-208VolumetricVolumetric50 mlEach$596.14Pack of 3$513.31Supplier Number: 70952-50
- 100489-196GraduatedGraduated2 mlEach$280.94Pack of 6$219.00Supplier Number: 70951-02
- 100489-200VolumetricVolumetric1 mlEach$278.76Pack of 6$239.50Supplier Number: 70952-01
- 100489-202VolumetricVolumetric5 mlEach$317.56Pack of 3$242.58Supplier Number: 70952-05
- 100489-206VolumetricVolumetric25 mlEach$385.32Pack of 3$294.31Supplier Number: 70952-25
- 100501-216VolumetricVolumetric2 mlEach$287.68Pack of 6$219.00Supplier Number: 70952-02
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.