Orders LinkContactUsLinkComponent
LEO SEM Mount, Electron Microscopy Sciences
LEO SEM Mount, Electron Microscopy Sciences
Certificates
LEO SEM Mount, Electron Microscopy Sciences
LEO SEM Mount, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These mounts with tapered end pin
  • Slotted head 12.7 mm (1/2") diameter
  • Pin 3.1 x 5 mm (1/8 x 1/4")
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 100488-660
    LEO Microscope Mounts Slotted Head
    LEO Microscope Mounts Slotted Head
    50
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $84.02
    $84.02
    Log in for availability
    Pack of 50
    Pack of 50 - $70.61
    $70.61
    Each
       $84.02
    Pack of 50
       $70.61
    Supplier Number: 75190
  • 100488-662
    LEO Microscope Mounts Slotted Head
    LEO Microscope Mounts Slotted Head
    100
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $172.94
    $172.94
    Log in for availability
    Pack of 100
    Pack of 100 - $144.73
    $144.73
    Each
       $172.94
    Pack of 100
       $144.73
    Supplier Number: 75191
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.