Orders LinkContactUsLinkComponent
Absorbant Paper Points, Electron Microscopy Sciences
Absorbant Paper Points, Electron Microscopy Sciences
Absorbant Paper Points, Electron Microscopy Sciences
Certificates
Absorbant Paper Points, Electron Microscopy Sciences
Absorbant Paper Points, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Machine rolled paper with a tapered end which offers good water absorbency, and assists you in the removal of surplus water from the knife boat, on the block face, and from the grids. Available in two assortment sizes. The bigger the number the larger the point.
Product Family Options
Product Information
  • Availability
    Price
  • 100500-760
    Paper Points, Selection 1
    Paper Points, Selection 1
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 200
    Box of 200 - $70.12
    $70.12
    Log in for availability
    Each
    Each - $70.12
    $70.12
    Box of 200
       $70.12
    Each
       $70.12
    Supplier Number: 71011-01
  • 100500-762
    Paper Points, Selection 2
    Paper Points, Selection 2
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 200
    Box of 200 - $58.31
    $58.31
    Log in for availability
    Each
    Each - $58.31
    $58.31
    Box of 200
       $58.31
    Each
       $58.31
    Supplier Number: 71011-02
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.