Five-Channel Timer, Electron Microscopy Sciences
About this item
Timer has four independent channels, each of which counts up or down for any interval from 1 second to 100 hours.
- Memory recalls last timer setting for reuse
- Has a clock mode
Its extra large display indicates channels that are in use. Each one has a distinctive 1 minute alarm. Automatically begins to count up to indicate time elapsed after count down reaches zero.
Dimensions: 2⁷/₈×2¹/₂×⁹/₁₆"
Weight: 2 oz.
Delivery information: Comes complete with a stand, pocket clip, and magnet.
2 Options Available Below
Product Details & Documents
Timer has four independent channels, each of which counts up or down for any interval from 1 second to 100 hours.
- Memory recalls last timer setting for reuse
- Has a clock mode
Its extra large display indicates channels that are in use. Each one has a distinctive 1 minute alarm. Automatically begins to count up to indicate time elapsed after count down reaches zero.
Dimensions: 2⁷/₈×2¹/₂×⁹/₁₆"
Weight: 2 oz.
Delivery information: Comes complete with a stand, pocket clip, and magnet.
Product Family Options
Product Information
- AvailabilityPrice
- 100493-814Timer 5-channel certifiedSupplier Number: 72600-02
- 100501-348Timer, 5-channelSupplier Number: 72600
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)