Reinforced Plastic Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Reinforced plastic tweezers made of PBTP (Polybutylenterephthalate) with 20% fiber-glass.
- Good resistance to chemicals and acids
- Withstand temperature upto 200 °C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)