Axygen® GEN3 Multi-Barrier Filter Tips, Sterile, Corning
Catalog #: 89167-688
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:1 - 20 µL
- Tip Color:Clear
- Sterile:+
- Supplier No.:GEN3-20LRS
Specifications
About this item
The new Axygen brand GEN3 multi-barrier filter pipette tips offer the ultimate in protection against cross contamination through tri-filter technology.
- Offered in racks and sterile
- Certified human gDNA and PCR inhibition free, DNase-/RNase-free, and nonpyrogenic
- Prevents liquid or aerosol from passing through and will lock on contact
GEN3 tips effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material. In addition to the self-sealing properties, GEN3 multi-barrier pipette tips feature the exclusive Maxymum Recovery® technology, which reduces the amount of reagent or sample adhering to the pipette tips.
Customers Who Bought This Also Bought