Order Entry
Canada
ContactUsLinkComponent
E.Z.N.A. PX Blood RNA Kit, Omega Bio-tek
E.Z.N.A. PX Blood RNA Kit, Omega Bio-tek
Catalog # CA89134-792
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A. PX Blood RNA Kit, Omega Bio-tek
Catalog # CA89134-792
Supplier:  Omega Bio-Tek
Supplier Number:  R1057-00
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Blood DNA/RNA collection tubes (e.g. PAXgene )
  • Target molecule:
    RNA
  • Description:
    E.Z.N.A. PX Blood RNA Kit
  • Size:
    5 Preps
  • Description:
    E.Z.N.A. PX Blood RNA Kit
  • Cat. no.:
    CA89134-792
  • Supplier No.:
    R1057-00

Specifications

About this item

Omega Bio-tek’s E.Z.N.A.® PX Blood RNA Kit is designed for isolation of total RNA from blood samples stored in special preserved reagents and Paxgene tubes

The procedure completely removes contaminants and enzyme inhibitors making RNA isolation fast, convenient, and reliable. RNA purified using the E.Z.N.A.® PX RNA Kit method is ready for applications such as RT-PCR.