Order Entry
Canada
ContactUsLinkComponent
SP Bel-Art® Bellows Samplers, Bel-Art Products, a part of SP
SP Bel-Art® Bellows Samplers, Bel-Art Products, a part of SP
Catalog # 13197-432
undefined
SP Bel-Art® Bellows Samplers, Bel-Art Products, a part of SP
Catalog # 13197-432
Supplier Number:  H37823-0050

Specifications

  • Capacity:
    50 ml
  • Length:
    50 cm (19½”)
  • Cat. no.:
    13197-432
  • Supplier No.:
    378230050

Specifications

About this item

Bellows samplers provide quick and easy liquid sampling with one-handed operation.

  • Polyethylene construction
  • Long, tapered tip
  • Quick and easy sampling
  • One-handed operation

The FDA-grade polyethylene sampler tube has a long tapered tip with a 4 mm opening.