ContactUsLinkComponent
SP Bel-Art® Three Zone Powder Sampler, Bel-Art Products, a part of SP
  70680-272
undefined
SP Bel-Art® Three Zone Powder Sampler, Bel-Art Products, a part of SP
  70680-272
 :  H37829-0000
  • Length:
    56 cm (22”)
  • Cat. no.:
    70680-272
  • Supplier No.:
    378290000
For proper sampling of powders, pellets and other friable materials which may become non-homogeneous in bags, this three zone sampler is invaluable.
  • Anodized aluminum construction with stainless steel point
  • Sampler is easy to use
  • Keeps individual samples homogeneous
Made of 1.9 cm (¾”) diameter anodized aluminum with a sharp stainless steel point; it has three ports 7.6 cm (3”) apart
The “T” handle is turned to close the sample cells, and the steel point is inserted through the bag wall. When the sampler is in deep enough to cover the three cells, a half turn of the handle allows samples to enter. The handle is then returned to the original position. The sampler is removed and the three samples transferred into containers for analysis.
The ports are 7.6 cm (3”) long x 1 cm (0.39”) wide with a volume of 9cc. The distance from the point to the first port is 4.3 cm (1¾”).