ContactUsLinkComponent
SP Bel-Art Curved Forceps, Bel-Art Products, a part of SP
SP Bel-Art Curved Forceps, Bel-Art Products, a part of SP
Catalog # 13197-442
undefined
SP Bel-Art Curved Forceps, Bel-Art Products, a part of SP
Catalog # 13197-442
Supplier Number:  H37940-0000
Specifications
  • Description:
    Curved Forceps
  • Environmentally Preferable:
  • Cat. no.:
    13197-442
  • Supplier No.:
    379400000
Specifications
About this item
The exceptionally fine tips of these forceps are coated with brown Fluo-Kem® Teflon® FEP which complies with FDA regulations covering direct contact with food.
  • FEP coating
  • Serrated, curved tips
  • Suited for grasping fine objects
They have an overall length of 9.8 cm (3⅞") and serrated, curved tips.