Tungsten Lamp for Lambda Spectrometers, PerkinElmer
Tungsten Lamp
Catalog # CA97012-474
Supplier: PerkinElmer
Supplier #: B0114620
Jump to:
Product Details & Documents
PerkinElmer’s long-lifetime deuterium and tungsten source lamps provide wavelength coverage across the working range of the spectrometer for consistently high performance.
- Long lifetime
- Broad range of wavelength coverage
- Consistently high performance
- Pre-aligned for easy replacement
All lamps are pre-aligned and can be easily changed by the user. Eye protection required when viewing deuterium or tungsten lamps when lit.
Ordering information: Adapter (P/N B0166354) may be required for older models of Lambda 5, 7 and 9.
Specifications
- Description:Tungsten Lamp