Order Entry
Canada
ContactUsLinkComponent
Mag-Bind® Viral DNA/RNA 96 Kit, Omega Bio-tek
Mag-Bind® Viral DNA/RNA 96 Kit, Omega Bio-tek
Catalog # CA76414-002
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind® Viral DNA/RNA 96 Kit, Omega Bio-tek
Catalog # CA76414-002
Supplier:  Omega Bio-Tek
Supplier Number:  M6246-03
CAS Number:  

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Viral RNA
    Viral DNA
  • Sample type:
    Swabs
    Blood
    Biofluids (serum, plasma, saliva)
  • Target molecule:
    Copurification of DNA & RNA
    DNA
    RNA
  • Description:
    Mag-Bind® Viral DNA/RNA 96 kit (12×96)
  • Size:
    12 x 96 Well Plate
  • Sample size:
    50 - 200 μl
  • Cat. no.:
    CA76414-002

Specifications

About this item

Mag-Bind® Viral DNA/RNA Kit is designed for the rapid and reliable isolation of viral RNA and viral DNA from whole Blood, serum, plasma, saliva and other body fluids

The Mag-Bind® magnetic beads technology enables purification of high-quality nucleic acids that is free of proteins, nucleases, and other impurities. In addition to easily being adapted with automated systems, this procedure can also be scaled up or down, allowing for the purification from various amounts of starting materials. The purified nucleic acid is ready for direct use in downstream applications, such as amplification or other enzymatic reactions.