Order Entry
Canada
ContactUsLinkComponent
Mag-Bind® Bacterial DNA 96 Kit, Omega Bio-tek
Mag-Bind® Bacterial DNA 96 Kit, Omega Bio-tek
Supplier:  Omega Bio-Tek
Certificates
Mag-Bind® Bacterial DNA 96 Kit, Omega Bio-tek
Supplier:  Omega Bio-Tek

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Swabs
    Bacteria
    Biofluids (serum, plasma, saliva)
  • Target molecule:
    DNA
  • Description:
    Mag-Bind® Bacterial DNA 96 Kit

Specifications

Related Information

Frequently Bought Together