Order Entry
Canada
ContactUsLinkComponent
E.Z.N.A.® M13 DNA Mini Kit, Omega Bio-tek
E.Z.N.A.® M13 DNA Mini Kit, Omega Bio-tek
Supplier:  Omega Bio-Tek
Certificates
E.Z.N.A.® M13 DNA Mini Kit, Omega Bio-tek
Supplier:  Omega Bio-Tek

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    ssDNA
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Description:
    E.Z.N.A.® M13 DNA Mini Kit

Specifications

Related Information

Frequently Bought Together