Chlorine Dioxide Color Disc Test Kit, Model CLD-2, Hach
: Hach
Monitor ClO2 levels on-site with the NEW Chlorine Dioxide Test Kit. The Chlorine Dioxide Test Kit uses the reliable and EPA-accepted DPD method to monitor ClO2 concentration, while eliminating chlorine interference through the addition of glycine. With 100 tests per kit and available range of 0-2 mg/L and resolution of 0.1 mg/L, operators can accurately monitor around the 0.8 mg/L regulated limit.
- Simple, cost-effective method for maintaining optimum chlorine dioxide levels in your plant
- USEPA-accepted for waste water and potable water
- Approved by some states for potable water under specific conditions (check with your regulatory agency)
: Kit includes two reagents, tube, color comparator box, color discs, instruction sheets, and carrying case.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Environmentally Preferable:
Frequently Bought Together