Tin Test Method: colorimetric with test strips and reagent 10 - 25 - 50 - 100 - 200 mg/L Sn MQuant™
Supplier: MilliporeSigma
Certificates
About this item
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot NumberEMD Millipore Simply Rapid MQuant and MColorpHast Mobile Analysis System MilliporeSigma Food and Beverage Applications
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together