

Ethanol absolute ≥99.5% (by GC), EMPARTA® ACS analytical reagent, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
Boiling Pt:78 °C (lit.)
Melting Pt:–114 °C (lit.)
Density:0.789 g/ml (25°C) (lit.)
Storage Temperature:Ambient
UN:1170
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
Specifications
Specification Test Results
- Solubility:(20 °C) completely miscible
- Melting Point:-114.5 °C
- Molar Mass:46.07 g/mol
- Boiling Point:78.3 °C (1013 hPa)
- Vapor Pressure:59 hPa (20 °C)
- Flash Point:12 °C
- Refractive Index:1.36
- Explosion Limit:3.5 - 15 %(V)
- Density:0.790 - 0.793 g/cm3 (20 °C)
- pH:7.0 (10 g/l, H2O, 20 °C)
- Ignition Point:425 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberEMD Millipore HDPE Bottles Flyer EMD Millipore Sustainable Products Brochure MilliporeSigma EasyGrip Full Control Ergonomic HDPE Bottlesfor Acids, Bases and Solvents MilliporeSigma Industrial Applications
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together