Alcian blue solution used in nuclear staining, for histology, Sigma-Aldrich®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:33864-99-2
:75881-23-1
Formula:C₅₆H₆₈Cl₄CuN₁₆S₄
MDL Number:MFCD00010720
:1298.86 g/mol
Density:1.006 g/cm3 (20 °C)
Storage Temperature:Ambient
:1
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Suitability for microscopy:passes test
- neutral mucopolysaccharides:purple
- polysaccharides:purple
- PAS negative acidic mucosubstances:light blue
- Nuclei:Blue
Specification Test Results
Related Information
Certificates
Search by Lot NumberMilliporeSigma Histology Tissue Diagnostics Workflow
Product Literature
Frequently Bought Together